Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Slovakia
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Standards
Organic Standards
Pesticide reference standard, (±)-α-BHC 100 µg/ml in n-nonane, CIL
Pesticide reference standard, (±)-α-BHC 100 µg/ml in n-nonane, CIL
Pesticide reference standard, (±)-α-BHC 100 µg/ml in n-nonane, CIL
PROOCILULM72321.2
:
PROMOCHEM
:
Pesticide reference standard, (±)-α-BHC 100 µg/ml in n-nonane, CIL
PROOCILULM72321.2
:
PROMOCHEM
:
Pack type:
Glass bottle
Pk:
1,2 mL
CAS No.:
319-84-6
Description:
(±)-α-BHC 100 µg/ml in n-nonane
Formula:
C₆H₆Cl₆
Description:
(±)-α-BHC