Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Slovakia
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Nitric acid
Nitric acid 1%, VOLUSOL® volumetric solution
Nitric acid 1%, VOLUSOL® volumetric solution
Nitric acid 1%, VOLUSOL® volumetric solution
VWRS83996.310
:
VWR Collection
:
Nitric acid 1%, VOLUSOL® volumetric solution
VWRS83996.310
:
VWR Collection
:
Pack type:
Glass bottle
Pk:
2,5 L
Seq:
5096
:
:
63.01 g/mol
:
Ambient
:
231-714-2
:
2031
:
8,II
:
GHS07
:
7697-37-2
:
HNO₃
:
MFCD00011349