Supelco® PESTANAL® Organic Reference Standard, Fenthion
36552-250MG
: Merck
:
- Pack type:Glass bottle for solids
- Pk:250 mg
- Standards:PESTANAL®, Analytical standard
- CAS No.:55-38-9
- Description:Fenthion
- Storage conditions:2...8 °C
- Hazard class description:H301 + H311 - H330 - H341 - H372 - H410
- Shipping conditions:STD
- MDL No.:MFCD00055449
- M-Clarity quality level:MQ100
- Synonym(s):O,O′-Dimethyl O″-[3-methyl-4-(methylthio)phenyl] thiophosphate
- Molecular weight:278,33
- Flash point:Not applicable
- GHS signal: GHS06, GHS08, GHS09
: P202 - P260 - P273 - P280 - P302 + P352 + P312 - P304 + P340 + P310