Supelco® PESTANAL® Organic Reference Standard, Cadusafos
Catalog # 32505-50MG
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:50 mg
- Standards:PESTANAL®, Analytical standard
- CAS No.:95465-99-9
- Description:Cadusafos
- Storage conditions:2...8 °C
- Hazard class description:H300 + H310 + H330 - H400
- Shipping conditions:STD
- MDL No.:MFCD00072511
- M-Clarity quality level:MQ100
- Synonym(s):O-Ethyl S,S-bis(1-methylpropyl) phosphorodithioate
- Molecular weight:270,39
- Flash point:129,4 °C
Specifications
About this item
- GHS signal: GHS06, GHS09
Caution: P262 - P264 - P273 - P280 - P302 + P352 + P310 - P304 + P340 + P310