Supelco® PESTANAL® Organic Reference Standard, Monocrotophos
36173-100MG
: Merck
:
- Pack type:Glass bottle for solids
- Pk:100 mg
- Standards:PESTANAL®, Analytical standard
- CAS No.:6923-22-4
- Description:Monocrotophos
- Storage conditions:2...8 °C
- Hazard class description:H300 + H330 - H311 - H341 - H410
- Shipping conditions:STD
- MDL No.:MFCD00055291
- M-Clarity quality level:MQ100
- Molecular weight:223,16
- Flash point:Not applicable
- GHS signal: GHS06, GHS08, GHS09
: P202 - P260 - P273 - P280 - P302 + P352 + P312 - P304 + P340 + P310