Order Entry
Slovakia
ContactUsLinkComponent
 
Supelco® PESTANAL® Organic Reference Standard, Ethalfluralin
  45472-250MG
 :  Merck
 :  
undefined
Supelco® PESTANAL® Organic Reference Standard, Ethalfluralin
  45472-250MG
 :  Merck
 :  

 

  • Pack type:
    Glass bottle for solids
  • Pk:
    250 mg
  • Standards:
    PESTANAL®, Analytical standard
  • CAS No.:
    55283-68-6
  • Description:
    Ethalfluralin
  • Storage conditions:
    2...30 °C
  • Hazard class description:
    H332 - H351 - H410
  • Shipping conditions:
    STD
  • MDL No.:
    MFCD00078681
  • M-Clarity quality level:
    MQ100
  • Molecular weight:
    333,26
  • Flash point:
    Not applicable

 

 

  • GHS signal: GHS07, GHS09
 : P202 - P261 - P271 - P273 - P304 + P340 + P312 - P308 + P313