Supelco® PESTANAL® Organic Reference Standard, Triflumuron
Catalog # 35029-100MG
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:100 mg
- Standards:PESTANAL®, Analytical standard
- CAS No.:64628-44-0
- Description:Triflumuron
- Storage conditions:2...30 °C
- Hazard class description:H330
- Shipping conditions:STD
- MDL No.:MFCD00072496
- M-Clarity quality level:MQ100
- Molecular weight:358,70
- Flash point:Not applicable
Specifications
About this item
- GHS signal: GHS06
Caution: P260 - P271 - P284 - P304 + P340 + P310 - P403 + P233 - P405