Supelco® PESTANAL® Organic Reference Standard, Carbaryl
32055-250MG
: Merck
:
- Pack type:Glass bottle for solids
- Pk:250 mg
- Standards:PESTANAL®, Analytical standard
- CAS No.:63-25-2
- Description:Carbaryl
- Storage conditions:2...30 °C
- Hazard class description:H302 + H332 - H351 - H400
- Shipping conditions:STD
- MDL No.:MFCD00021467
- M-Clarity quality level:MQ100
- Synonym(s):1-Naphthyl-N-methylcarbamate
- Melting point:142...146 °C (lit.)
- Formula:C₁₀H₇OCONHCH₃
- Molecular weight:201,22
- Flash point:Not applicable
- GHS signal: GHS06, GHS08, GHS09
: P202 - P261 - P273 - P301 + P312 - P304 + P340 + P312 - P308 + P313