Supelco® PESTANAL® Organic Reference Standard, Chlorflurenol
45721-100MG
: Merck
:
- Pk:100 mg
- Standards:PESTANAL®, analytical standard
- CAS No.:2464-37-1
- Description:Chlorflurenol
- Storage conditions:2...30 °C
- Shipping conditions:STD
- MDL No.:MFCD00056359
- M-Clarity quality level:MQ100
- Synonym(s):2-Chloro-9-hydroxyflurene-9-carboxylic acid
- Molecular weight:260,67