Supelco® VETRANAL® Organic Reference Standard, Arecoline Hydrobromide
31593-250MG
: Merck
:
- Pack type:Glass bottle for solids
- Pk:250 mg
- Standards:VETRANAL®, Analytical standard
- CAS No.:300-08-3
- Description:Arecoline hydrobromide
- Storage conditions:2...30 °C
- Hazard class description:H302
- Shipping conditions:STD
- MDL No.:MFCD00039041
- M-Clarity quality level:MQ100
- Synonym(s):1-Methyl-1,2,5,6-tetrahydro-3-pyridinecarboxylic acid methyl ester hydrobromide; Arecaidine methyl ester hydrobromide; Methyl 1,2,5,6-tetrahydro-1-methyl-3-pyridinecarboxylate hydrobromide; Methyl 1-methyl-1,2,5,6-tetrahydronicotinate hydrobromide; Taeniolin
- Melting point:172...176 °C (lit.)
- Molecular weight:236,11
- Flash point:Not applicable
- GHS signal: GHS07
: P264 - P270 - P301 + P312 - P501