Order Entry
Slovakia
ContactUsLinkComponent
Supelco® PESTANAL® Organic Reference Standard, 2,4,5-Trichlorophenoxyacetic acid
Supelco® PESTANAL® Organic Reference Standard, 2,4,5-Trichlorophenoxyacetic acid
Catalog # 45667-250MG
Supplier:  Merck
CAS Number:  
undefined
Supelco® PESTANAL® Organic Reference Standard, 2,4,5-Trichlorophenoxyacetic acid
Catalog # 45667-250MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    250 mg
  • Standards:
    PESTANAL®
  • CAS No.:
    93-76-5
  • Description:
    2,4,5-T
  • Storage conditions:
    2...30 °C
  • Hazard class description:
    H301 - H312 - H315 - H319 - H335 - H410
  • Shipping conditions:
    STD
  • MDL No.:
    MFCD00004301
  • M-Clarity quality level:
    MQ100
  • Synonym(s):
    2,4,5-Trichlorophenoxyacetic acid; 2,4,5-T
  • Melting point:
    154...158 °C (lit.)
  • Formula:
    Cl₃C₆H₂OCH₂CO₂H
  • Molecular weight:
    255,48
  • Flash point:
    Not applicable

Specifications

About this item

  • GHS signal: GHS07, GHS09
Caution: P261 - P273 - P280 - P301 + P310 - P302 + P352 + P312 - P305 + P351 + P338