Order Entry
Slovakia
ContactUsLinkComponent
Silicon standard solution, 1,000 mg/L Si in 0.5 M sodium hydroxide solution (from SiO2) standard for AAS
Silicon standard solution, 1,000 mg/L Si in 0.5 M sodium hydroxide solution (from SiO2) standard for AAS
Catalog # SCARSI00110100
Supplier:  SCHARLAB
CAS Number:  
Silicon standard solution, 1,000 mg/L Si in 0.5 M sodium hydroxide solution (from SiO2) standard for AAS
Catalog # SCARSI00110100
Supplier:  SCHARLAB
CAS Number:  

Specifications

  • Concentration:
    1000 mg/L
  • Matrix:
    0,5 M NaOH
  • Pack type:
    Glass bottle
  • Pk:
    100 ml
  • Element:
    Silicon
  • Symbol:
    Si

Specifications

About this item