



n-Heptane for HPLC
Catalog # SCARHE01312500
Supplier: SCHARLAB
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:2,5 L
- Seq:5096
Specifications
About this item
:
:100.20 g/mol
Merck Index:13,04679
Boiling Pt:98.4 °C (1013 hPa)
Melting Pt:–90.6 °C
Density:0.6838 g/cm³ (20 °C)
Flash Pt:–4 °C
Storage Temperature:Ambient
EINECS:205-563-8
UN:1206
ADR:3,II
:GHS02
:GHS08
:GHS07
:GHS09
CAS Number:142-82-5
Formula:H₃C(CH₂)₅CH₃
MDL Number:MFCD00009544