Supelco® PESTANAL® Organic Reference Standard, Aldrin
Catalog # 36666-25MG
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 mg
- Standards:PESTANAL®, Analytical standard
- CAS No.:309-00-2
- Description:Aldrin
- Storage conditions:2...30 °C
- Hazard class description:H300 + H310 - H351 - H372 - H410
- Shipping conditions:STD
- MDL No.:MFCD00135587
- M-Clarity quality level:MQ100
- Molecular weight:364,91
- Flash point:Not applicable
Specifications
About this item
- GHS signal: GHS06, GHS08, GHS09
Caution: P202 - P264 - P273 - P280 - P301 + P310 - P302 + P352 + P310