Order Entry
Slovakia
ContactUsLinkComponent
Manganese standard solution, 1000 mg/L Mn in 0.5 M nitric acid (from Mn(NO₃)₂) standard for AAS
Manganese standard solution, 1000 mg/L Mn in 0.5 M nitric acid (from Mn(NO₃)₂) standard for AAS
Catalog # SCARMA01110500
Supplier:  SCHARLAB
CAS Number:  
Manganese standard solution, 1000 mg/L Mn in 0.5 M nitric acid (from Mn(NO₃)₂) standard for AAS
Catalog # SCARMA01110500
Supplier:  SCHARLAB
CAS Number:  

Specifications

  • Concentration:
    1000 mg/L
  • Matrix:
    0,5 M HNO₃
  • Pack type:
    Glass bottle
  • Pk:
    500 mL
  • Element:
    Manganese
  • Symbol:
    Mn

Specifications

About this item