Sodium standard solution, 1000 mg/L Na in 0,5 M nitric acid (from NaNO₃) standard for AAS
Catalog # SCARSO00050500
Supplier: SCHARLAB
CAS Number:
Specifications
- Concentration:1000 mg/L
- Matrix:0,5 M HNO₃
- Pack type:Glass bottle
- Pk:500 mL
- Element:Sodium
- Symbol:Na