Molybdenum standard solution, 1000 mg/L Mo in water (from (NH₄)₆Mo₇O₂₄) standard for AAS
Catalog # SCARMO00210100
Supplier: SCHARLAB
CAS Number:
Specifications
- Concentration:1000 mg/L
- Matrix:H₂O
- Pack type:Glass bottle
- Pk:1 L
- Element:Molybdenum
- Symbol:Mo