Order Entry
Slovakia
ContactUsLinkComponent
2,2,6-Trimethyl-4H-1,3-dioxin-4-one, Sigma-Aldrich®
2,2,6-Trimethyl-4H-1,3-dioxin-4-one, Sigma-Aldrich®
Catalog # SIAL245100-100G
Supplier:  Merck
CAS Number:  
2,2,6-Trimethyl-4H-1,3-dioxin-4-one, Sigma-Aldrich®
Catalog # SIAL245100-100G
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g
  • Seq:
    3401

Specifications

About this item

 :
 :142.15 g/mol
 :M-Clarity™ quality level = MQ200
EINECS:226-403-3
CAS Number:5394-63-8
Formula:C₇H₁₀O₃
MDL Number:MFCD00040468