

2,2,6-Trimethyl-4H-1,3-dioxin-4-one ≥94% cont. up to ca 6% acetone
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:5394-63-8
:
Formula:C₇H₁₀O₃
MDL Number:MFCD00040468
:142.15 g/mol
Boiling Pt:65…67 °C (2 mmHg)
Melting Pt:7 °C
Density:1.094
Flash Pt:14 °C (57 °F)
EINECS:226-403-3
ADR:3,II
:GHS02
:GHS07
Related Information
Frequently Bought Together