
Sodium mercaptoacetate (sodium thioglycolate), Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:367-51-1
:
Formula:HSCH₂COONa
MDL Number:MFCD00043386
:114.1 g/mol
:M-Clarity™ quality level = MQ200
EINECS:206-696-4
:GHS07
Storage Temperature:Freezer
Frequently Bought Together