
2-(Methylthio)ethanol 99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:5271-38-5
:
Formula:C₃H₈OS
MDL Number:MFCD00002908
Density:1.06 g/cm³
Flash Pt:70 °C
Storage Temperature:Ambient
EINECS:226-090-3
UN:3334
ADR:9,III
:92.16 g/mol
:GHS07
Specification Test Results
- Appearance:Clear colorless Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4934 to 1.4954 (20°C, 589 nm)
- GC:≥98.5 %
Specification Test Results
Frequently Bought Together