Arsenic standard solution, 1000 mg/L As in 0.5 M hydrochloric acid (from As₂O₃) standard for AAS
Catalog # SCARAR01510100
Supplier: SCHARLAB
CAS Number:
Specifications
- Concentration:1000 mg/L
- Matrix:0,5 M HNO₃
- Pack type:Glass bottle
- Pk:100 mL
- Element:Arsenic
- Symbol:As