
Sodium hydroxide, pellets, Reagent Grade
Catalog # SCARSO04250500
Supplier: SCHARLAB
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:500 g
- Seq:3645
Specifications
About this item
:
:40 g/mol
Merck Index:14,08627
Boiling Pt:1390 °C (1013 hPa)
Melting Pt:323 °C
Density:2.13 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:215-185-5
UN:1823
ADR:8,II
REACH:01-2119457892-27
:GHS05
CAS Number:1310-73-2
Formula:NaOH
MDL Number:MFCD00003548