Aluminium standard solution, 1000 mg/L Al in 0,5 M nitric acid (from Al(NO₃)₃.9H₂O) standard for AAS
Catalog # SCARAL07510500
Supplier: SCHARLAB
CAS Number:
Specifications
- Concentration:1000 mg/L
- Matrix:0,5 M HNO₃
- Pack type:Glass bottle
- Pk:500 mL
- Element:Aluminium
- Symbol:Al