Tris(2-ethylhexyl)phosphate, Supelco®
Catalog # 93299-25ML
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:25 mL
- Seq:2315
Specifications
About this item
:
:434.64 g/mol
:M-Clarity™ quality level = MQ100
EINECS:201-116-6
CAS Number:78-42-2
Formula:C₂₄H₅₁O₄P
MDL Number:MFCD00009491