Order Entry
Slovakia
ContactUsLinkComponent
Tris(2-ethylhexyl) phosphate
Tris(2-ethylhexyl) phosphate
Catalog # EHERC17894400
Supplier:  EHRENSTORFER
CAS Number:  
Tris(2-ethylhexyl) phosphate
Catalog # EHERC17894400
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    0,25 g
  • Seq:
    3029

Specifications

About this item

 :
 :434.64 g/mol
EINECS:201-116-6
CAS Number:78-42-2
Formula:C₂₄H₅₁O₄P
MDL Number:MFCD00009491