Order Entry
Slovakia
Orders LinkContactUsLinkComponent
Naphthalene, extra pure

Naphthalene, extra pure

Glass ampoule, 100 mg
Catalog # 123092C
Health
Irritant
Environment
Supplier:   VWR Chemicals
Naphthalene, extra pure
Catalog # 123092C
Supplier:  VWR Chemicals
CAS Number:  91-20-3
Naphthalene, extra pure
Naphthalene, extra pure

Glass ampoule, 100 mg

CAS Number:  91-20-3
Price
44,80 €
Log in for availability
1 * 100 mg
Product Details & Documents
  • CAS Number:91-20-3
  • Formula:C₁₀H₈
  • MDL Number:MFCD00001742
  • Molecular Weight:128.17 g/mol
  • Merck Index:12,06457
  • Boiling Pt:218 °C (1013 hPa)
  • Melting Pt:79…82 °C
  • Density:1.145 g/cm³ (20 °C)
  • Flash Pt:79 °C
  • Storage Temperature:Ambient
  • EINECS:202-049-5
  • UN:1334
  • ADR:4.1,III
Specifications

Specifications

  • Pack type:
    Glass ampoule
  • Pk:
    100 mg

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.