Naphthalene ≥99.6%
About this item
Purity: 99.6% CAS No.: 91-20-3 Molecular Formula: C10H8 Formula Weight: 128.17 Flash Point: 78°(172°F) Physical Form: Crystalline Density: 0.9628 Refractive Index: 1.5821 Odour: Aromatic Melting Point: 80-82° Boiling Point: 218° Solubility: Soluble in benzene, toluene, chloroform, CS2, ether. Insoluble in water Beilstein Reference: 1421310 Merck Reference: 14,6370 UN No.: 1334 MDL No.: MFCD00001742
Formerly important in dyestuff manufacture. As a precursor to phthalic anhydride
- CAS Number:91-20-3
- Formula:C₁₀H₈
- MDL Number:MFCD00001742
- Molecular Weight:128.17 g/mol
- Merck Index:14,6370
- Boiling Pt:218 °C
- Melting Pt:80…82 °C
- Density:0.9628
- Flash Pt:78 °C (172 °F)
- Storage Temperature:Ambient
- EINECS:202-049-5
- UN:1334
- ADR:4.1,III
3 Options Available Below
Product Details & Documents
Purity: 99.6% CAS No.: 91-20-3 Molecular Formula: C10H8 Formula Weight: 128.17 Flash Point: 78°(172°F) Physical Form: Crystalline Density: 0.9628 Refractive Index: 1.5821 Odour: Aromatic Melting Point: 80-82° Boiling Point: 218° Solubility: Soluble in benzene, toluene, chloroform, CS2, ether. Insoluble in water Beilstein Reference: 1421310 Merck Reference: 14,6370 UN No.: 1334 MDL No.: MFCD00001742
Formerly important in dyestuff manufacture. As a precursor to phthalic anhydride
- CAS Number:91-20-3
- Formula:C₁₀H₈
- MDL Number:MFCD00001742
- Molecular Weight:128.17 g/mol
- Merck Index:14,6370
- Boiling Pt:218 °C
- Melting Pt:80…82 °C
- Density:0.9628
- Flash Pt:78 °C (172 °F)
- Storage Temperature:Ambient
- EINECS:202-049-5
- UN:1334
- ADR:4.1,III
Product Family Options
Product Information
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



