Order Entry
Slovakia
ContactUsLinkComponent
 
 
 
Barium nitrate ≥99.0%, AnalaR NORMAPUR® analytical reagent
  21740.293
 :  
undefined
 
 
Barium nitrate ≥99.0%, AnalaR NORMAPUR® analytical reagent
  21740.293
 :  

 

  • Pack type:
    Plastic bottle for solids
  • Pk:
    1 kg
  • Seq:
    4024

 

 

  • Assay:
    Min. 99.0 %
  • pH (20°C; 5 %):
    5.0 - 8.0
  • Heavy metals (as Pb):
    Max. 5 ppm
  • Insolubility in water:
    Max. 100 ppm
  • Not precipitated by H2SO4 (as SO4):
    Max. 0.05 %
  • Cl (Chloride):
    Max. 5 ppm
  • Ca (Calcium):
    Max. 20 ppm
  • Fe (Iron):
    Max. 2 ppm
  • K (Potassium):
    Max. 50 ppm
  • Na (Sodium):
    Max. 50 ppm
  • Sr (Strontium):
    Max. 0.2 %

 

 

 :
 :13,00986
 :∼592 °C
 :∼3.23 g/cm³ (20 °C)
 :5.1 (6.1),II
 :261.34 g/mol
 :233-020-5
 :1446
 :GHS03
 :GHS07
 :Ambient
 :10022-31-8
 :Ba(NO₃)₂
 :MFCD00003442