
Calcium chloride
Catalog # ABCRAV16337.1000
Supplier: abcr
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 kg
Specifications
About this item
:
Merck Index:14,01659
Boiling Pt:1600 °C (1013 hPa)
Melting Pt:772 °C
Density:2.174 g/cm³ (20 °C)
:110.98 g/mol
Storage Temperature:Ambient
EINECS:233-140-8
REACH:01-2119494219-28
:GHS07
CAS Number:10043-52-4
Formula:CaCl₂
MDL Number:MFCD00010903