
Barium chloride, ultra dry ≥99.998% (metals basis)
Catalog # 13686.14
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:208.24 g/mol
Merck Index:14,971
Boiling Pt:1560 °C
Melting Pt:963 °C
Density:3.9
Storage Temperature:Ambient
EINECS:233-788-1
UN:1564
ADR:6.1,III
:ampouled under argon
:GHS06
CAS Number:10361-37-2
Formula:BaCl₂
MDL Number:MFCD00003445