Sodium orthovanadate 99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:13721-39-6
:
Formula:Na₃O₄V
MDL Number:MFCD00003511
:183.91 g/mol
Melting Pt:850…866 °C
EINECS:237-287-9
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:White to off-white Powder
- Assay:≥98.5 % (as Na3VO4)
- Aluminium (Al):≤200 ppm
- Iron (Fe):≤30 ppm
- Potassium (K):≤150 ppm
- Silicon (Si):≤200 ppm
- Vanadium (V):26.5 % (typically)
Specification Test Results
Frequently Bought Together