Order Entry
Slovakia
ContactUsLinkComponent
Sodium peroxodisulphate ≥99.0%, AnalaR NORMAPUR® analytical reagent
Oxidizing
Irritant
Health
Sodium peroxodisulphate ≥99.0%, AnalaR NORMAPUR® analytical reagent
Supplier:  VWR Chemicals
Certificates
undefined
Oxidizing
Irritant
Health
Sodium peroxodisulphate ≥99.0%, AnalaR NORMAPUR® analytical reagent
Supplier:  VWR Chemicals
Certificates

Specification Test Results

  • Assay:
    Min. 99.0 %
  • Acidity or alkalinity:
    Max. 0.01 meq/g
  • Heavy metals (as Pb):
    Max. 10 ppm
  • Insolubility in water:
    Max. 50 ppm
  • Cl (Chloride):
    Max. 5 ppm
  • NH4 (Ammonium):
    Max. 100 ppm
  • Fe (Iron):
    Max. 10 ppm
  • Mn (Manganese):
    Max. 2 ppm

Specification Test Results

Frequently Bought Together