Order Entry
Slovakia
ContactUsLinkComponent
 
Supelco® Organic Reference Standard, Naphthalene-d₈ solution
  48715-U
 :  Merck
 :  
Supelco® Organic Reference Standard, Naphthalene-d₈ solution
  48715-U
 :  Merck
 :  

 

  • Concentration:
    2000 μg/ml in dichloromethane
  • Pk:
    1
  • Standards:
    Certified reference material
  • CAS No.:
    1146-65-2
  • Description:
    Naphthalene-d₈ solution
  • Storage conditions:
    2…8 °C
  • Hazard class description:
    H315 - H319 - H336 - H351 - H373 - H373
  • Shipping conditions:
    STD
  • M-Clarity quality level:
    MQ100

 

 

  • GHS signal: GHS08, GHS07
 : P201 - P202 - P260 - P302 + P352 - P305 + P351 + P338 - P308 + P313