Supelco® Organic Reference Standard, Didecyl Phthalate
36933-250MG
: Merck
:
- Pack type:Glass bottle for solids
- Pk:250 mg
- Standards:Analytical standard
- CAS No.:84-77-5
- Description:Didecyl phthalate
- Storage conditions:2...30 °C
- Shipping conditions:STD
- MDL No.:MFCD00041915
- M-Clarity quality level:MQ100
- Formula:C₆H₄[COO(CH₂)₉CH₃]₂
- Molecular weight:446,66
- Flash point:200 °C - closed cup