Order Entry
Slovakia
ContactUsLinkComponent
Supelco® Organic Reference Standard, (−)-α-Pinene
Supelco® Organic Reference Standard, (−)-α-Pinene
Supplier:  Merck
Certificates
undefined
Supelco® Organic Reference Standard, (−)-α-Pinene
Supplier:  Merck

Specifications

  • CAS No.:
    7785-26-4
  • Storage conditions:
    2…8 °C
  • Hazard class description:
    H226 - H302 - H304 - H315 - H410
  • Shipping conditions:
    STD
  • MDL No.:
    MFCD00064145
  • M-Clarity quality level:
    MQ100
  • Synonym(s):
    (1S,5S)-2,6,6-Trimethylbicyclo[3.1.1]hept-2-ene; (1S,5S)-2-Pinene
  • Molecular weight:
    136,23
  • Density:
    0,858 g/ml at 20 °C (lit.)
  • Flash point:
    31,3 °C
  • Boiling point:
    156 - 158 °C (lit.)

Specifications

Frequently Bought Together