

m-Xylene, Supelco®
Catalog # 95670-5ML
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:5 mL
- Seq:2210
Specifications
About this item
:
:106.17 g/mol
Merck Index:12,10214
Boiling Pt:138 °C (1013 hPa)
Melting Pt:–48 °C
Density:0.864 g/cm³ (20 °C)
Flash Pt:25 °C
EINECS:203-576-3
UN:1307
ADR:3,III
:M-Clarity™ quality level = MQ100
:GHS02
:GHS07
Storage Temperature:Ambient
CAS Number:108-38-3
Formula:C₈H₁₀
MDL Number:MFCD00008536