Kovac's indole reagent, Millipore®
Catalog # SIAL67309-100ML-F
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:100 mL
- Seq:2440
Specifications
About this item
:
:149.19 g/mol
Merck Index:13,03257
Boiling Pt:176…177 °C (1013 hPa)
Melting Pt:72 °C
Density:1.0254 g/cm³ (99.9 °C)
Flash Pt:>100 °C
Storage Temperature:Ambient
EINECS:202-819-0
CAS Number:100-10-7
Formula:(CH₃)₂NC₆H₄CHO
MDL Number:MFCD00003381