Order Entry
Slovakia
Orders LinkContactUsLinkComponent
Methanol

Methanol

Flammable
Toxic
Health
   Sustainable Options Available
Supplier:   SCHARLAB
Methanol
Supplier:  SCHARLAB
   Sustainable Options Available
Certificates
Methanol

2 Options Available Below

Product Details & Documents
Synonyms:  Wood alcohol, Methyl alcohol, Hydroxy methane, Carbinol
  • CAS Number:67-56-1
  • Formula:CH₃OH
  • MDL Number:MFCD00004595
  • Molecular Weight:32.04 g/mol
  • Merck Index:13,05984
  • Boiling Pt:64.6 °C (1013 hPa)
  • Melting Pt:–98 °C
  • Density:0.7918 g/cm³ (20 °C)
  • Flash Pt:11 °C (closed cup)
  • Storage Temperature:Ambient
  • EINECS:200-659-6
  • UN:1230
  • ADR:3,II
  • REACH:01-2119433307-44
Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • SCARME03261000
  • SCARME03262500
  • Environmentally Preferable:
  • Safer Human and Environmental Health
  • Safer Human and Environmental Health
  • Environmental certification:
  • Chem21 Solvent Selection Guide recommended after discussion
  • Chem21 Solvent Selection Guide recommended after discussion

Specifications

Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle
    1 L
    Log in for availability
    Log in for availability
    1 * 1 L
    1 * 1 L

    Specifications:
      Safer Human and Environmental Health
    More
    Specifications
    Cat. No.
    SCARME03261000
    Environmentally Preferable
    Safer Human and Environmental Health
    Environmental certification
    Chem21 Solvent Selection Guide recommended after discussion
      Safer Human and Environmental Health
  • Glass bottle
    2,5 L
    Log in for availability
    Log in for availability
    1 * 2,5 L
    1 * 2,5 L

    Specifications:
      Safer Human and Environmental Health
    More
    Specifications
    Cat. No.
    SCARME03262500
    Environmentally Preferable
    Safer Human and Environmental Health
    Environmental certification
    Chem21 Solvent Selection Guide recommended after discussion
      Safer Human and Environmental Health
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.