Methanol
Supplier: SCHARLAB
Jump to:
About this item
Synonyms: Wood alcohol, Methyl alcohol, Hydroxy methane, Carbinol
- CAS Number:67-56-1
- Formula:CH₃OH
- MDL Number:MFCD00004595
- Molecular Weight:32.04 g/mol
- Merck Index:13,05984
- Boiling Pt:64.6 °C (1013 hPa)
- Melting Pt:–98 °C
- Density:0.7918 g/cm³ (20 °C)
- Flash Pt:11 °C (closed cup)
- Storage Temperature:Ambient
- EINECS:200-659-6
- UN:1230
- ADR:3,II
- REACH:01-2119433307-44
2 Options Available Below
Product Details & Documents
Synonyms: Wood alcohol, Methyl alcohol, Hydroxy methane, Carbinol
- CAS Number:67-56-1
- Formula:CH₃OH
- MDL Number:MFCD00004595
- Molecular Weight:32.04 g/mol
- Merck Index:13,05984
- Boiling Pt:64.6 °C (1013 hPa)
- Melting Pt:–98 °C
- Density:0.7918 g/cm³ (20 °C)
- Flash Pt:11 °C (closed cup)
- Storage Temperature:Ambient
- EINECS:200-659-6
- UN:1230
- ADR:3,II
- REACH:01-2119433307-44
Product Family Options
Product Information
- Pack typePkAvailabilityPrice



