α-Benzoinoxime ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:441-38-3
:
Formula:C₁₄H₁₃NO₂
MDL Number:MFCD00004501
:227.26 g/mol
Merck Index:14,1094
Melting Pt:151…154 °C
Storage Temperature:Ambient
EINECS:207-127-2
Related Information
Frequently Bought Together