3-Methylbiphenyl 95%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:643-93-6
:
Formula:C₁₃H₁₂
MDL Number:MFCD00008533
:168.24 g/mol
Boiling Pt:272 °C
Melting Pt:4…5 °C
Density:1.01 g/cm³
Flash Pt:>110 °C
Storage Temperature:Ambient
EINECS:211-404-3
Specification Test Results
- Appearance:Clear light yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.6010 to 1.6050 (20°C, 589 nm)
- GC:≥94 %
Specification Test Results
Frequently Bought Together