



Propylbenzene, Supelco®
Supplier: Merck
Certificates
About this item
CAS Number:103-65-1
:
Formula:C₉H₁₂
MDL Number:MFCD00009377
:120.19 g/mol
Merck Index:12,08029
Boiling Pt:159 °C (1013 hPa)
Melting Pt:–99 °C
Density:0.862 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:203-132-9
UN:2364
ADR:3,III
:M-Clarity™ quality level = MQ100
Flash Pt:45 °C
:GHS02
:GHS08
:GHS07
:GHS09
Frequently Bought Together