Supelco® Organic Reference Standard, Toluene
Supplier: Merck
Certificates
About this item
- GHS signal: GHS02, GHS08, GHS07
Caution: P202 - P210 - P273 - P301 + P310 - P303 + P361 + P353 - P331
Specifications
- CAS No.:108-88-3
- Storage conditions:2...30 °C
- Hazard class description:H225 - H304 - H315 - H336 - H361d - H373 - H412
- Shipping conditions:STD
- MDL No.:MFCD00008512
- M-Clarity quality level:MQ100
- Melting point:–93 °C (lit.)
- Formula:C₆H₅CH₃
- Molecular weight:92,14
- Density:0,865 g/ml at 25 °C (lit.)
- Flash point:4,0 °C
- Boiling point:110 - 111 °C (lit.)
Specifications
Frequently Bought Together