Dodecylbenzene for synthesis, Sigma-Aldrich®
: Merck
:123-01-3
:
:C₁₈H₃₀
:MFCD00008974
:275 °C (1013 hPa)
:–4 °C
:0.85 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ200
:246.44 g/mol
:Ambient
:204-591-8
- Solubility:(20 °C) insoluble
- Melting Point:3 °C
- Molar Mass:246.44 g/mol
- Boiling Point:275 - 300 °C (1013 hPa)
- Vapor Pressure:0.1 hPa (50 °C)
- Flash Point:124 °C
- Refractive Index:1.482 (20 °C, 589 nm)
- Density:0.85 g/cm3 (20 °C)
- Synonyms:Dodecyl benzene
- Hill Formula:C18H30
- Chemical Formula:C6H5(CH2)11CH3
Frequently Bought Together