Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Slovakia
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
4-Isopropylphenol
4-Isopropylphenol
4-Isopropylphenol
EHERC14464100
:
EHRENSTORFER
:
4-Isopropylphenol
EHERC14464100
:
EHRENSTORFER
:
Pack type:
Glass bottle for solids
Pk:
0,25 g
Seq:
3029
:
:
136.19 g/mol
:
213 °C (1013 hPa)
:
60 °C
:
0.99 g/cm³ (20 °C)
:
Ambient
:
202-798-8
:
2430
:
8,III
:
GHS05
:
99-89-8
:
C₉H₁₂O
:
MFCD00002372