Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Slovakia
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
7-Benzyloxygramine
7-Benzyloxygramine, Supelco®
7-Benzyloxygramine, Supelco®
7-Benzyloxygramine, Supelco®
Catalog # B9875-100MG
Supplier:
Merck
CAS Number:
Expand image
7-Benzyloxygramine, Supelco®
Catalog # B9875-100MG
Supplier:
Merck
CAS Number:
Specifications
Pack type:
Glass bottle for solids
Pk:
100 mg
Seq:
2440
Specifications
About this item
Product Details
:
:
280.37 g/mol
CAS Number:
94067-27-3
Formula:
C₁₈H₂₀N₂O
Product Details