4-Nitrophenol ≥99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:100-02-7
:
Formula:C₆H₅NO₃
MDL Number:MFCD00007331
:139.11 g/mol
Merck Index:14,6621
Boiling Pt:279 °C (dec.)
Melting Pt:110…115 °C
Density:1.480
Flash Pt:169 °C (336 °F)
Storage Temperature:Ambient
EINECS:202-811-7
UN:1663
ADR:6.1,III
:GHS08
:GHS07
Related Information
Frequently Bought Together