Fluorobenzene, Supelco®
Catalog # 51411-5ML-F
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:5 mL
- Seq:2210
Specifications
About this item
:
:96.1 g/mol
Merck Index:13,04197
Boiling Pt:85.1 °C (1013 hPa)
Melting Pt:–41 °C
Density:1.023 g/cm³ (20 °C)
Flash Pt:–15 °C
Storage Temperature:Ambient
EINECS:207-321-7
UN:2387
ADR:3,II
:M-Clarity™ quality level = MQ100
CAS Number:462-06-6
Formula:C₆H₅F
MDL Number:MFCD00000280