Order Entry
Slovakia
ContactUsLinkComponent
Calcium nitrate tetrahydrate
Irritant
Corrosive
Calcium nitrate tetrahydrate
Catalog # VWRS6078.9025
Supplier:  VWR Collection
CAS Number:  
Irritant
Corrosive
Calcium nitrate tetrahydrate
Catalog # VWRS6078.9025
Supplier:  VWR Collection
CAS Number:  

Specifications

  • Pk:
    25 kg
  • Seq:
    4283

Specifications

About this item

 :
 :236.15 g/mol
Merck Index:13,01687
Melting Pt:∼42 °C
Density:1.86 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:233-332-1
UN:1454
ADR:5.1,III
 :GHS07
 :GHS05
CAS Number:13477-34-4
Formula:Ca(NO₃)₂·4H₂O
MDL Number:MFCD00149604